6-Fluoronicotinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoro-5-pyridinecarboxylic acid
H55147-03
H55147-MD
AAH55147-03EA
146
USD
AAH55147-03
AAH55147-MD
6-Fluoronicotinic acid 97%
6-Fluoronicotinic acid
Purity: 97%
CAS No.: 403-45-2
Molecular Formula: C6H4FNO2
Formula Weight: 141.10
Melting Point: 144-148°
MDL No.: MFCD01859863
Formula:
C₆H₄FNO₂ MW: 141.1 g/mol Melting Pt: 144…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD01859863 CAS Number: 403-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...