Silver(I) acetylacetonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Silver 2,4-pentanedionate, Silver acetylacetonate
H55131-03
H55131-06
H55131-14
AAH55131-03EA
68.58
USD
AAH55131-03
AAH55131-06
AAH55131-14
Silver(I) acetylacetonate 98%
Silver(I) acetylacetonate
Purity: 98%
CAS No.: 15525-64-1
Molecular Formula: C5H7AgO2
Formula Weight: 206.98
Storage Temperature: 2°C to 8°C
Odor: Characteristic
Melting Point: 100° dec.
MDL No.: MFCD00000012
Formula:
C₅H₈AgO₂ MW: 207.985 g/mol Melting Pt: 100 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00000012 CAS Number: 15525-64-1 EINECS: 239-568-1 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...