1-Benzyl N-Carbobenzoxy-D-glutamate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-Benzyl N-Cbz-D-glutamate, Z-D-Glu-OBzl, N-CBZ-D-glutamic acid alpha-benzyl ester, N-Carbobenzoxy-D-glutamic Acid 1-Benzyl Ester, N-Cbz-D-glutamic Acid 1-Benzyl Ester, Z-D-Glu-Obzl
H55121-03
H55121-06
AAH55121-03EA
60
USD
AAH55121-03
AAH55121-06
1-Benzyl N-Carbobenzoxy-D-glutamate 97%
1-Benzyl-N-carbobenzoxy-D-glutamate
Purity: 97%
CAS No.: 65706-99-2
Molecular Formula: C20H21NO6
Formula Weight: 371.39
Beilstein Reference: 5305848
MDL No.: MFCD00069647
Formula:
C₂₀H₂₁NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00069647 CAS Number: 65706-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...