Monoethyl succinate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl hydrogen succinate, 4-Ethoxy-4-oxobutanoic acid, mono-Ethyl succinate

H55116-03 H55116-06 H55116-14
AAH55116-03EA 29.6 USD
AAH55116-03 AAH55116-06 AAH55116-14
Monoethyl succinate 95%
Monoethyl succinate

Purity: 95% CAS No.: 1070-34-4 Molecular Formula: C6H10O4 Formula Weight: 146.14 Flash Point: 113°(235°F) Refractive Index: 1.4320 Boiling Point: 248° MDL No.: MFCD00053346

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 248 °C
Density: 1.141
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053346
CAS Number: 1070-34-4
EINECS: 213-973-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR