6-Bromoquinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

H55111-03 H55111-06
AAH55111-03EA 45 USD
AAH55111-03 AAH55111-06 AAH55111-MD
6-Bromoquinoline 97%
6-Bromoquinoline

Purity: 97% CAS No.: 5332-25-2 Molecular Formula: C9H6BrN Formula Weight: 208.06 Flash Point: 110°(230°F) Refractive Index: 1.6630 Melting Point: 24° MDL No.: MFCD00024023

Formula: C₉H₆BrN
MW: 208.06 g/mol
Melting Pt: 24 °C
Density: 1.538
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00024023
CAS Number: 5332-25-2
EINECS: 226-238-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR