Ethyl-2-bromothiazole-5-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-bromothiazole-5-carboxylate

H55109-03 H55109-06
AAH55109-03EA 69.2 USD
AAH55109-03 AAH55109-06 AAH55109-MD
Ethyl-2-bromothiazole-5-carboxylate 98%
Ethyl-2-bromothiazole-5-carboxylate

Purity: 98% CAS No.: 41731-83-3 Molecular Formula: C6H6BrNO2S Formula Weight: 236.08 Flash Point: 110°(230°F) Refractive Index: 1.5530 Boiling Point: 216-217° MDL No.: MFCD00463837

Formula: C₆H₆BrNO₂S
Boiling Pt: 216…217 °C
Density: 1.581
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00463837
CAS Number: 41731-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR