3-Ethyl-4-methyl-3-pyrrolin-2-one 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one, 1,5-Dihydro-3-ethyl-4-methyl-2H-pyrrol-2-one

H55065-03 H55065-06
AAH55065-03EA 34.2 USD
AAH55065-03 AAH55065-06 AAH55065-MD
3-Ethyl-4-methyl-3-pyrrolin-2-one 98%
3-Ethyl-4-methyl-3-pyrrolin-2-one

Purity: 98% CAS No.: 766-36-9 Molecular Formula: C7H11NO Formula Weight: 125.17 Melting Point: 100-104° MDL No.: MFCD00173861

Formula: C₇H₁₁NO
MW: 125.17 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00173861
CAS Number: 766-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR