1,4-Bis(2-hydroxyethyl)piperazine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2'-(Piperazine-1,4-diyl)diethanol
H55063-06
H55063-14
AAH55063-06EA
46.3
USD
AAH55063-06
AAH55063-14
1,4-Bis(2-hydroxyethyl)piperazine 98%
1,4-Bis(2-hydroxyethyl)piperazine
Purity: 99%
CAS No.: 122-96-3
Molecular Formula: C8H18N2O2
Formula Weight: 174.24
Melting Point: 133-136°
Boiling Point: 215-220°/50mm
MDL No.: MFCD00006157
Formula:
C₈H₁₈N₂O₂ MW: 174.24 g/mol Boiling Pt: 215…220 °C/50 mm Melting Pt: 133…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006157 CAS Number: 122-96-3 EINECS: 204-586-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...