4-[(Trimethylsilyl)ethynyl]benzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-((Trimethylsilyl)ethynyl)benzaldehyde, 4-(Trimethylsilylethynyl)benzaldehyde
H55060-03
H55060-06
H55060-14
AAH55060-03EA
54.2
USD
AAH55060-03
AAH55060-06
AAH55060-14
4-[(Trimethylsilyl)ethynyl]benzaldehyde 97%
4-[(Trimethylsilyl)ethynyl]benzaldehyde
Purity: 97%
CAS No.: 77123-57-0
Molecular Formula: C12H14OSi
Formula Weight: 202.33
Melting Point: 66-70°
MDL No.: MFCD02093765
Formula:
C₁₂H₁₄OSi MW: 202.33 g/mol Melting Pt: 66…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093765 CAS Number: 77123-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...