tert-Butyl-3-hydroxyphenylcarbamate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl (3-hydroxyphenyl)carbamate
H55059-03
H55059-06
H55059-14
AAH55059-03EA
22.9
USD
AAH55059-03
AAH55059-06
AAH55059-14
tert-Butyl-3-hydroxyphenylcarbamate 97%
tert-Butyl-3-hydroxyphenylcarbamate
Purity: 97%
CAS No.: 19962-06-2
Molecular Formula: C11H15NO3
Formula Weight: 209.25
Melting Point: 134-138°
MDL No.: MFCD06798070
Formula:
C₁₁H₁₅NO₃ MW: 209.25 g/mol Melting Pt: 134…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD06798070 CAS Number: 19962-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...