2,2-Bis[4-(4-aminophenoxy)phenyl]propane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4'-Isopropylidenebis[(4-aminophenoxy)benzene], 4,4'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))dianiline

H55057-14 H55057-22
AAH55057-14EA 46.7 USD
AAH55057-14 AAH55057-22
2,2-Bis[4-(4-aminophenoxy)phenyl]propane 98%
2,2-Bis[4-(4-aminophenoxy)phenyl]propane

Purity: 98% CAS No.: 13080-86-9 Molecular Formula: C27H26N2O2 Formula Weight: 410.52 Melting Point: 127-130° MDL No.: MFCD00039152

Formula: C₂₇H₂₆N₂O₂
MW: 410.52 g/mol
Melting Pt: 127…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00039152
CAS Number: 13080-86-9
EINECS: 235-985-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR