S-Phenyl-L-cysteine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-2-Amino-3-(phenylthio)propanoic acid, H-Cys(phenyl)-OH

H55031-03 H55031-06 H55031-14
AAH55031-03EA 32 USD
AAH55031-03 AAH55031-06 AAH55031-14
S-Phenyl-L-cysteine 97%
S-Phenyl-L-cysteine

Purity: 97% CAS No.: 34317-61-8 Molecular Formula: C9H11NO2S Formula Weight: 197.25 Melting Point: 200° dec. Beilstein Reference: 2268203 Optical Rotation: +10° (c=1.5 in 1M NaOH) MDL No.: MFCD01318758

Formula: C₉H₁₁NO₂S
MW: 197.252 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01318758
CAS Number: 34317-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR