3-[(Trimethylsilyl)ethynyl]benzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-((Trimethylsilyl)ethynyl)benzonitrile
H55012-03
H55012-06
AAH55012-03EA
59.8
USD
AAH55012-03
AAH55012-06
AAH55012-MD
3-[(Trimethylsilyl)ethynyl]benzonitrile 97%
3-[(Trimethylsilyl)ethynyl]benzonitrile
Purity: 97%
CAS No.: 190771-22-3
Molecular Formula: C12H13NSi
Formula Weight: 199.33
Melting Point: 42-47°
MDL No.: MFCD09260494
Formula:
C₁₂H₁₃NSi MW: 199.33 g/mol Melting Pt: 42…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD09260494 CAS Number: 190771-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...