Indole-7-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
1H-Indole-7-carboxylic acid
H54931-03
H54931-06
H54931-14
AAH54931-03EA
42.7
USD
AAH54931-03
AAH54931-06
AAH54931-14
Indole-7-carboxylic acid 97%
Indole-7-carboxylic acid
Purity: 97%
CAS No.: 1670-83-3
Molecular Formula: C9H7NO2
Formula Weight: 161.05
MDL No.: MFCD00210442
Formula:
C₉H₇NO₂ MW: 161.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210442 CAS Number: 1670-83-3 EINECS: 216-801-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...