(S)-(-)-2-Amino-2-methyl-4-pentenoic acid ≥98%, ee 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54922-03
H54922-MD
AAH54922-03EA
456
USD
AAH54922-03
AAH54922-MD
(S)-(-)-2-Amino-2-methyl-4-pentenoic acid ≥98%, ee 99%
(S)-(-)-2-Amino-2-methyl-4-pentenoic acid
Purity: 98%
CAS No.: 96886-55-4
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Storage Temperature: 2°C to 8°C
Melting Point: 285°
Optical Rotation: -14° (c=1.3 in 1N HCl)
MDL No.: MFCD00145248
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Melting Pt: 285 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145248 CAS Number: 96886-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...