5-Bromo-3-methoxyquinoline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54919-03
H54919-06
H54919-MD
AAH54919-03EA
188
USD
AAH54919-03
AAH54919-06
AAH54919-MD
5-Bromo-3-methoxyquinoline 96%
5-Bromo-3-methoxyquinoline
Purity: 96%
CAS No.: 776296-12-9
Molecular Formula: C10H8BrNO
Formula Weight: 238.08
MDL No.: MFCD16038652
Formula:
C₁₀H₈BrNO MW: 238.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16038652 CAS Number: 776296-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...