Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TFFH, N-((Dimethylamino)fluoromethylene)-N-methylmethanaminium hexafluorophosphate(V)

H54910-03 H54910-06
AAH54910-03EA 54.8 USD
AAH54910-03 AAH54910-06 AAH54910-14
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate 98%
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate

Purity: 98% CAS No.: 164298-23-1 Molecular Formula: C5H12F7N2P Formula Weight: 264.12 Melting Point: 104-111° MDL No.: MFCD02684443

Formula: C₅H₁₂F₇N₂P
MW: 264.13 g/mol
Melting Pt: 104…111 °C
Storage Temperature: Ambient
MDL Number: MFCD02684443
CAS Number: 164298-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR