6-Bromo-2-chloroquinoline 96%

Supplier: Thermo Scientific Chemicals
Warning

H54895-03 H54895-06
AAH54895-03EA 119 USD
AAH54895-03 AAH54895-06 AAH54895-MD
6-Bromo-2-chloroquinoline 96%
6-Bromo-2-chloroquinoline

Purity: 96% CAS No.: 1810-71-5 Molecular Formula: C9H5BrClN Formula Weight: 242.50 Storage Temperature: 2°C to 8°C Melting Point: 148-150° MDL No.: MFCD04115272

Formula: C₉H₅BrClN
MW: 242.50 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD04115272
CAS Number: 1810-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR