Bis(pentafluorophenyl)carbonate 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Bis(pentafluorophenyl) Carbonate
H54880-06
H54880-14
AAH54880-03EA
50.25
USD
AAH54880-03
AAH54880-06
AAH54880-14
Bis(pentafluorophenyl)carbonate 98+%
Bis(pentafluorophenyl)carbonate
Purity: 98+%
CAS No.: 59483-84-0
Molecular Formula: C13F10O3
Formula Weight: 394.12
Flash Point: 110°(230°F)
Melting Point: 47-50°
Beilstein Reference: 2029727
MDL No.: MFCD00368353
Formula:
C₁₃F₁₀O₃ MW: 394.13 Melting Pt: 47…50 °C Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00368353 CAS Number: 59483-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...