4-[(2,4-Dimethoxyphenyl)-(fmoc-amino)methyl]phenoxyacetic acid ≥98%

Supplier: Thermo Scientific Chemicals

H54874-03 H54874-06
AAH54874-03EA 148 USD
AAH54874-03 AAH54874-06
4-[(2,4-Dimethoxyphenyl)-(fmoc-amino)methyl]phenoxyacetic acid ≥98%
4-[(2,4-Dimethoxyphenyl)-(fmoc-amino)methyl]phenoxyacetic acid

Purity: 98+% CAS No.: 145069-56-3 Molecular Formula: C32H29NO7 Formula Weight: 539.58 Storage Temperature: 2°C to 8°C Melting Point: ca 180° dec. MDL No.: MFCD00153509

Formula: C₃₂H₂₉NO₇
Melting Pt: ca 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00153509
CAS Number: 126828-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR