Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium 1-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carbonyl)oxy)-2,5-dioxopyrrolidine-3-sulphonate, Sulphosuccinimidyl-trans-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, Sulpho-SMCC Sodium Salt, SMCC, Water-Soluble, 4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt, sulpho-SMCC, Sulfo-SMCC
H54807-MB
H54807-MC
AAH54807-MBEA
166
USD
AAH54807-MB
AAH54807-MC
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥97%
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Purity: 98+%
CAS No.: 92921-24-9
Molecular Formula: C16H17N2NaO9S
Formula Weight: 436.37
Storage Temperature: 2°C to 8°C
MDL No.: MFCD00054978
Formula:
C₁₆H₁₈N₂O₉S MW: 436.37 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00054978 CAS Number: 92921-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...