2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine
H54793-03
H54793-MD
AAH54793-03EA
314
USD
AAH54793-03
AAH54793-MD
2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 96%
2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester
Purity: 96%
CAS No.: 947249-01-6
Molecular Formula: C12H16BF3N2O2
Formula Weight: 288.07
Melting Point: 132-134°
MDL No.: MFCD12923414
Formula:
C₁₂H₁₆BF₃N₂O₂ MW: 288.07 g/mol Melting Pt: 132…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD12923414 CAS Number: 947249-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...