1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole ≥99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(Mesitylsulphonyl)-3-nitro-1H-1,2,4-triazole

H54792-03 H54792-06
AAH54792-03EA 96.9 USD
AAH54792-03 AAH54792-06
1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole ≥99%
1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole

Purity: 99+% CAS No.: 74257-00-4 Molecular Formula: C11H12N4O4S Formula Weight: 296.30 Melting Point: 134-139° Beilstein Reference: 4328921 MDL No.: MFCD00009754

Formula: C₁₁H₁₂N₄O₄S
MW: 296.301 g/mol
Melting Pt: 134…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00009754
CAS Number: 74257-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR