2-Amino-3-methylpyridine-5-boronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
H54773-03
H54773-MD
AAH54773-03EA
308
USD
AAH54773-03
AAH54773-MD
2-Amino-3-methylpyridine-5-boronic acid pinacol ester 96%
2-Amino-3-methylpyridine-5-boronic acid pinacol ester
Purity: 96%
CAS No.: 1111637-91-2
Molecular Formula: C12H19BN2O2
Formula Weight: 234.10
Storage Temperature: 2°C to 8°C
Melting Point: 128-130°
MDL No.: MFCD12923388
Formula:
C₁₂H₁₉BN₂O₂ Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD12923388 CAS Number: 1111637-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...