7-Bromo-1-heptanol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54762-03
H54762-06
AAH54762-03EA
81.52
USD
AAH54762-03
AAH54762-06
AAH54762-MD
7-Bromo-1-heptanol 96%
7-Bromo-1-heptanol
Purity: 96%
CAS No.: 10160-24-4
Molecular Formula: C7H15BrO
Formula Weight: 195.10
Flash Point: 113°(235°F)
Refractive Index: 1.4820
Boiling Point: 111-112°/4mm
MDL No.: MFCD00041708
Formula:
C₇H₁₅BrO MW: 195.10 g/mol Boiling Pt: 111…112 °C/4 mm Density: 1.269 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041708 CAS Number: 10160-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...