O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥99%

Supplier: Thermo Scientific Chemicals
Warning

H54732-14 H54732-22
AAH54732-06EA 24.21 USD
AAH54732-06 AAH54732-14 AAH54732-22
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥99%
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate

Purity: 99+% CAS No.: 330641-16-2 Molecular Formula: C11H15BClF4N5O Formula Weight: 355.53 Melting Point: 198-204° MDL No.: MFCD04973270

Formula: C₁₁H₁₅BClF₄N₅O
Melting Pt: 198…204 °C
Storage Temperature: Ambient
MDL Number: MFCD04973270
CAS Number: 330641-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR