2-Cyano-5-pyridineboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile, 2-(6-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Cyano-3-pyridylboronic acid Pinacol ester, 2-Cyanopyridine-5-boronic acid pinacol ester

H54689-03 H54689-MD
AAH54689-03EA 272 USD
AAH54689-03 AAH54689-MD
2-Cyano-5-pyridineboronic acid pinacol ester 96%
2-Cyano-5-pyridineboronic acid pinacol ester

Purity: 96% CAS No.: 741709-63-7 Molecular Formula: C12H15BN2O2 Formula Weight: 230.07 Melting Point: 105-106° MDL No.: MFCD06657825

Formula: C₁₂H₁₅BN₂O₂
Melting Pt: 105…106 °C
Storage Temperature: Ambient
MDL Number: MFCD06657825
CAS Number: 741709-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR