1-Chloroacetyl-2-(trifluoroacetyl)hydrazine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(Chloroacetyl)-2-(trifluoroacetyl)hydrazine

H54647-03 H54647-06 H54647-MD
AAH54647-03EA 126 USD
AAH54647-03 AAH54647-06 AAH54647-MD
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine 97%
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine

Purity: 97% CAS No.: 762240-99-3 Molecular Formula: C4H4ClF3N2O2 Formula Weight: 204.53 MDL No.: MFCD07779449

Formula: C₄H₄ClF₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07779449
CAS Number: 762240-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR