2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide

H54614-03 H54614-MD
AAH54614-03EA 336 USD
AAH54614-03 AAH54614-MD
2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%
2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester

Purity: 96% CAS No.: 1006876-27-2 Molecular Formula: C14H21BN2O3 Formula Weight: 276.14 MDL No.: MFCD08458484

Formula: C₁₄H₂₁BN₂O₃
MW: 276.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458484
CAS Number: 1006876-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR