tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis-Boc-pyrazolocarboxamidine, 1-[N,N'-Bis(tert-butoxycarbonyl)carbamimidoyl]-1H-pyrazole, N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, N,N'-Di-Boc-1H-pyrazole-1-carboximidamide, 1H-Pyrazole-1-carboxamidine, N,N'-Bis-BOC protected, N,N'-Bis(tert-butoxycarbonyl)-1H-pyrazole-1-carboximidamide

H54565-03 H54565-06 H54565-14
AAH54565-03EA 42.4 USD
AAH54565-03 AAH54565-06 AAH54565-14
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate

Purity: 98+% CAS No.: 152120-54-2 Molecular Formula: C14H22N4O4 Formula Weight: 310.35 Melting Point: 86-90° MDL No.: MFCD01075122

Formula: C₁₄H₂₂N₄O₄
MW: 310.354 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD01075122
CAS Number: 152120-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR