4,6-Difluoroindole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54536-03
H54536-06
AAH54536-03EA
92.2
USD
AAH54536-03
AAH54536-06
AAH54536-MD
4,6-Difluoroindole 97%
4,6-Difluoroindole
Purity: 97%
CAS No.: 199526-97-1
Molecular Formula: C8H5F2N
Formula Weight: 153.13
Storage Temperature: 2°C to 8°C
Formula:
C₈H₅F₂N MW: 153.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01075213 CAS Number: 199526-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...