Pentafluorophenyl trifluoroacetate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Trifluoroacetic Acid Pentafluorophenyl Ester, Pentafluorphenyl trifluoracetate
H54529-06
H54529-14
AAH54529-06EA
90
USD
AAH54529-06
AAH54529-14
Pentafluorophenyl trifluoroacetate ≥98%
Pentafluorophenyl trifluoroacetate
Purity: 98+%
CAS No.: 14533-84-7
Molecular Formula: C8F8O2
Formula Weight: 280.07
Flash Point: 52°(126°F)
Refractive Index: 1.368
Boiling Point: 122-123°
Beilstein Reference: 2003848
UN No.: 3272
MDL No.: MFCD00134438
Formula:
C₈F₈O₂ MW: 280.07 g/mol Boiling Pt: 122…123 °C Density: 1.63 Flash Pt: 52 °C (126 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00134438 CAS Number: 14533-84-7 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...