2-Phenyl-4-thiazolecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenylthiazole-4-carboxylic acid

H54521-03 H54521-06 H54521-MD
AAH54521-03EA 86.6 USD
AAH54521-03 AAH54521-06 AAH54521-MD
2-Phenyl-4-thiazolecarboxylic acid 97%
2-Phenyl-4-thiazolecarboxylic acid

Purity: 97% CAS No.: 7113-10-2 Molecular Formula: C10H7NO2S Formula Weight: 205.23 Melting Point: 148-152° MDL No.: MFCD00141954

Formula: C₁₀H₇NO₂S
MW: 205.23 g/mol
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00141954
CAS Number: 7113-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR