4-Fluoro-3-nitrobenzenesulfonamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H54516-03 H54516-MD
AAH54516-03EA 203 USD
AAH54516-03 AAH54516-MD
4-Fluoro-3-nitrobenzenesulfonamide 97%
4-Fluoro-3-nitrobenzenesulphonamide

Purity: 97% CAS No.: 406233-31-6 Molecular Formula: C6H5FN2O4S Formula Weight: 220.18 MDL No.: MFCD08703185

Formula: C₆H₅FN₂O₄S
MW: 220.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08703185
CAS Number: 406233-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR