1-(4-Bromophenyl)-N-methylmethanamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54506-03
H54506-06
AAH54506-03EA
50.4
USD
AAH54506-03
AAH54506-06
1-(4-Bromophenyl)-N-methylmethanamine 97%
1-(4-Bromophenyl)-N-methylmethanamine
Purity: 97%
CAS No.: 699-03-6
Molecular Formula: C8H10BrN
Formula Weight: 200.08
Flash Point:
Formula:
C₈H₁₀BrN MW: 200.08 g/mol Boiling Pt: 231…232 °C Density: 1.461 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04632143 CAS Number: 699-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...