N-(tert-Butoxycarbonyloxy)succinimide ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl (2,5-dioxopyrrolidin-1-yl) carbonate
H54493-03
H54493-06
H54493-14
AAH54493-03EA
31.4
USD
AAH54493-03
AAH54493-06
AAH54493-14
N-(tert-Butoxycarbonyloxy)succinimide ≥98%
N-(tert-Butoxycarbonyloxy)succinimide
Purity: 98+%
CAS No.: 13139-12-3
Molecular Formula: C9H13NO5
Formula Weight: 215.20
Storage Temperature: -30°C to -10°C
Melting Point: ca 95° dec.
MDL No.: MFCD00037903
Formula:
C₉H₁₃NO₅ MW: 215.20 g/mol Melting Pt: ∼ 95 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00037903 CAS Number: 13139-12-3 EINECS: 236-071-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...