4-Methyl-2-phenyl-1,3-thiazole-5-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methyl-2-phenylthiazole-5-carboxylic acid

H54487-03 H54487-06
AAH54487-03EA 58.4 USD
AAH54487-03 AAH54487-06 AAH54487-MD
4-Methyl-2-phenyl-1,3-thiazole-5-carboxylic acid 97%
4-Methyl-2-phenyl-1,3-thiazole-5-carboxylic acid

Purity: 97% CAS No.: 33763-20-1 Molecular Formula: C11H9NO2S Formula Weight: 219.26 Melting Point: 202-206° MDL No.: MFCD00141955

Formula: C₁₁H₉NO₂S
MW: 219.26 g/mol
Melting Pt: 202…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00141955
CAS Number: 33763-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR