Pyrimidine-5-carboxaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Pyrimidinecarboxaldehyde
H54448-03
H54448-06
H54448-14
AAH54448-03EA
82.6
USD
AAH54448-03
AAH54448-06
AAH54448-14
Pyrimidine-5-carboxaldehyde 97%
Pyrimidine-5-carboxaldehyde
Purity: 97%
CAS No.: 10070-92-5
Molecular Formula: C5H4N2O
Formula Weight: 108.10
Melting Point: 39-43°
Boiling Point: 80-82°/4mm
MDL No.: MFCD03426065
Formula:
C₅H₄N₂O MW: 108.1 g/mol Boiling Pt: 80…82 °C/4 mm Melting Pt: 39…43 °C Density: 1.23 Storage Temperature: Ambient |
MDL Number:
MFCD03426065 CAS Number: 10070-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...