2-Fluoro-N-methylbenzylamine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Fluorophenyl)-N-methylmethanamine, (2-Fluorobenzyl)methylamine

H54439-03 H54439-06
AAH54439-03EA 53.2 USD
AAH54439-03 AAH54439-06
2-Fluoro-N-methylbenzylamine 95%
2-Fluoro-N-methylbenzylamine

Purity: 95% CAS No.: 399-30-4 Molecular Formula: C8H10FN Formula Weight: 139.17 Flash Point: 68°(154°F) Physical Form: Liquid Density: 1.047 Refractive Index: 1.047 Sensitivity: Air Sensitive Appearance: Colorless to tan Boiling Point: 73° MDL No.: MFCD04629633

Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 73 °C
Density: 1.047
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD04629633
CAS Number: 399-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR