4-(1,1'-Biphenyl-4-yl)-1,3-thiazol-2-amine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-([1,1'-Biphenyl]-4-yl)thiazol-2-amine

H54413-03 H54413-06 H54413-MD
AAH54413-03EA 156 USD
AAH54413-03 AAH54413-06 AAH54413-MD
4-(1,1'-Biphenyl-4-yl)-1,3-thiazol-2-amine 97%
4-(1,1'-Biphenyl-4-yl)-1,3-thiazol-2-amine

Purity: 97% CAS No.: 2834-79-9 Molecular Formula: C15H12N2S Formula Weight: 252.33 Beilstein Reference: 187642 MDL No.: MFCD00047059

Formula: C₁₅H₁₂N₂S
MW: 252.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047059
CAS Number: 2834-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR