N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine ≥98%

Supplier: Thermo Scientific Chemicals
Warning

H54394-06 H54394-14
AAH54394-03EA 21.35 USD
AAH54394-03 AAH54394-06 AAH54394-14
N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine ≥98%
N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine

Purity: 98+% CAS No.: 152120-61-1 Molecular Formula: C9H14N4O2 Formula Weight: 210.23 Melting Point: 97-101° Beilstein Reference: 6141956 MDL No.: MFCD00216663

Formula: C₉H₁₄N₄O₂
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00216663
CAS Number: 152120-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR