Monomethyl terephthalate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl hydrogen terephthalate, 4-(Methoxycarbonyl)benzoic acid

H54391-06 H54391-14 H54391-22
AAH54391-06EA 45.5 USD
AAH54391-06 AAH54391-14 AAH54391-22
Monomethyl terephthalate 99+%
Monomethyl terephthalate

Purity: 99+% CAS No.: 1679-64-7 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: 220-223° Beilstein Reference: 1911082

Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 220…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00002557
CAS Number: 1679-64-7
EINECS: 216-849-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR