5-Cyano-3-pyridineboronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile, 3-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Cyanopyridine-5-boronic acid Pinacol ester, 3-Cyanopyridine-5-boronic acid pinacol ester, 2-(5-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H54385-03
H54385-MD
AAH54385-03EA
171
USD
AAH54385-03
AAH54385-MD
5-Cyano-3-pyridineboronic acid pinacol ester 96%
5-Cyano-3-pyridineboronic acid pinacol ester
Purity: 96%
CAS No.: 402718-29-0
Molecular Formula: C12H15BN2O2
Formula Weight: 230.07
Melting Point: 122-124°
MDL No.: MFCD07780755
Formula:
C₁₂H₁₅BN₂O₂ Melting Pt: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD07780755 CAS Number: 402718-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...