(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, (3-Hydroxy-3H-1,2,3-triazolo[4,5-b]pyridinato-O)tri-1-pyrrolidinylphosphonium hexafluorophosphate, ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), PyAOP, (3-Hydroxy-3H-1,2,3-triazolo[4,5-b]pyridinato-O)tri-1-pyrrolidinylphosphorous hexafluorophosphate

H54378-03 H54378-06
AAH54378-03EA 87.77 USD
AAH54378-03 AAH54378-06
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 99+%
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)

Purity: 99+% CAS No.: 156311-83-0 Molecular Formula: C17H27F6N7OP2 Formula Weight: 521.38 Melting Point: 163-168° MDL No.: MFCD03703417

Formula: C₁₇H₂₇F₆N₇OP₂
MW: 521.4 g/mol
Melting Pt: 163…168 °C
Storage Temperature: Ambient
MDL Number: MFCD03703417
CAS Number: 156311-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR