2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine, 2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester
H54372-03
H54372-MD
AAH54372-03EA
566
USD
AAH54372-03
AAH54372-MD
2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 95%
2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester
Purity: 95%
CAS No.: 944401-57-4
Molecular Formula: C12H16BF3N2O2
Formula Weight: 288.07
Beilstein Reference: 15519102
MDL No.: MFCD12923420
Formula:
C₁₂H₁₆BF₃N₂O₂ MW: 288.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12923420 CAS Number: 944401-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...