4-Fluoro-N-methylbenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Methyl-4-fluorobenzylamine

H54369-03 H54369-06
AAH54369-03EA 42 USD
AAH54369-03 AAH54369-06
4-Fluoro-N-methylbenzylamine 97%
4-Fluoro-N-methylbenzylamine

Purity: 97% CAS No.: 405-66-3 Molecular Formula: C8H10FN Formula Weight: 139.17 Flash Point: 71°(160°F) Refractive Index: 1.499 Boiling Point: 180-181° MDL No.: MFCD03839848

Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 180…181 °C
Density: 1.056
Flash Pt: 71 °C(160 °F)
Storage Temperature: Ambient
MDL Number: MFCD03839848
CAS Number: 405-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR