4-[4-(4'-Chloro-2-biphenylylmethyl)-1-piperazinyl]benzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54342-03
H54342-MD
AAH54342-03EA
163
USD
AAH54342-03
AAH54342-MD
4-[4-(4'-Chloro-2-biphenylylmethyl)-1-piperazinyl]benzoic acid 97%
4-[4-(4'-Chloro-2-biphenylylmethyl)-1-piperazinyl]benzoic acid
Purity: 97%
CAS No.: 916204-05-2
Molecular Formula: C24H23ClN2O2
Formula Weight: 406.91
MDL No.: MFCD16251290
Formula:
C₂₄H₂₃ClN₂O₂ MW: 406.91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16251290 CAS Number: 916204-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...