(1H-Indol-2-yl)methanamine 97%

Supplier: Thermo Scientific Chemicals
Warning

H54281-03 H54281-MD
AAH54281-03EA 229 USD
AAH54281-03 AAH54281-06 AAH54281-MD
(1H-Indol-2-yl)methanamine 97%
(1H-Indol-2-yl)methanamine

Purity: 97% CAS No.: 21109-25-1 Molecular Formula: C9H10N2 Formula Weight: 146.19 MDL No.: MFCD03422512

Formula: C₉H₁₀N₂
MW: 146.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03422512
CAS Number: 21109-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR