2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one 97%

Supplier: Thermo Scientific Chemicals

H54266-03 H54266-06
AAH54266-03EA 134 USD
AAH54266-03 AAH54266-06 AAH54266-MD
2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one 97%
2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one

Purity: 97% CAS No.: 3158-91-6 Molecular Formula: C13H8ClNO2 Formula Weight: 245.66 MDL No.: MFCD06658244

Formula: C₁₃H₈ClNO₂
MW: 245.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658244
CAS Number: 3158-91-6
EINECS: 221-602-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR