4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-iodoquinazolin-4-amine

H54229-03 H54229-MD
AAH54229-03EA 403 USD
AAH54229-03 AAH54229-MD
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline 97%
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline

Purity: 97% CAS No.: 231278-20-9 Molecular Formula: C21H14ClFIN3O Formula Weight: 505.72 MDL No.: MFCD09998827

Formula: C₂₁H₁₄ClFIN₃O
Storage Temperature: Ambient
MDL Number: MFCD09998827
CAS Number: 231278-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR